APO429356972 Apollo Scientific Ltd

2,4-Difluoro-5-nitrobenzyl alcohol

  • MDL number MFCD12047726
  • Cas Number : 935287-88-0
  • Molecular Weight : 189.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-difluoro-5-nitrophenyl)methanol
Inchi Code 1S/C7H5F2NO3/c8-5-2-6(9)7(10(12)13)1-4(5)3-11/h1-2,11H,3H2
Inchi Key VPJNXLNSNZEHSQ-UHFFFAOYSA-N

Description

Synonym (2,4-Difluoro-5-nitrophenyl)methanol
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluoro-5-nitrobenzyl-alcohol

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/24-difluoro-5-nitrobenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart