APO432343943 Apollo Scientific Ltd

Ethyl 2-ethoxy-2-iminoacetate

  • MDL number MFCD06797654
  • Cas Number : 816-27-3
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.042
Flash Point 72.4
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-ethoxy-2-iminoacetate

Safety Information

signalword Warning
pcodes P501
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-ethoxy-2-iminoacetate
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart