APO443483640 Apollo Scientific Ltd

Ethyl 2,5-dichlorothiophene-3-carboxylate

  • Cas Number : 130562-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=68974

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=68974
Availability details for
Quantity
  • Availability
  • Add to Cart