APO443483657 Apollo Scientific Ltd

3-Bromo-1H-indole-2-carboxylic acid

  • MDL number MFCD02710285
  • Cas Number : 28737-33-9
  • Molecular Weight : 240.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-1H-indole-2-carboxylic acid
Inchi Code 1S/C9H6BrNO2/c10-7-5-3-1-2-4-6(5)11-8(7)9(12)13/h1-4,11H,(H,12,13)
Inchi Key KOTZNHLNBDTYMS-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-1h-indole-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-1h-indole-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart