APO443483659 Apollo Scientific Ltd

1-Methyl-1H-indole-2-carbonitrile

  • MDL number MFCD09263865
  • Cas Number : 60680-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=69036

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=69036
Availability details for
Quantity
  • Availability
  • Add to Cart