APO443483856 Apollo Scientific Ltd

trans-3-Benzylamino-4-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD09608005
  • Cas Number : 138026-89-8
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3R,4R)-3-(benzylamino)-4-hydroxy-1-pyrrolidinecarboxylate
Inchi Code 1S/C16H24N2O3/c1-16(2,3)21-15(20)18-10-13(14(19)11-18)17-9-12-7-5-4-6-8-12/h4-8,13-14,17,19H,9-11H2,1-3H3/t13-,14-/m1/s1
Inchi Key GVYATPKTSSTHKN-ZIAGYGMSSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/trans-3-benzylamino-4-hydroxy-pyrrolidine-1-carboxylic-acid-tert-butyl-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/trans-3-benzylamino-4-hydroxy-pyrrolidine-1-carboxylic-acid-tert-butyl-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart