APO443485242 Apollo Scientific Ltd

6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD01717328
  • Cas Number : 57196-62-0
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 233-236
Iupac Name 6-methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Inchi Code 1S/C10H13NO.ClH/c1-12-10-3-2-9-7-11-5-4-8(9)6-10;/h2-3,6,11H,4-5,7H2,1H3;1H
Inchi Key QIUIKPQXMJJOQT-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 233-236
Product Link https://store.apolloscientific.co.uk/product/6-methoxy-1234-tetrahydroisoquinoline-hydrochloride

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/6-methoxy-1234-tetrahydroisoquinoline-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart