APO443485357 Apollo Scientific Ltd

Ethyl 4-(carbamoylamino)benzoate

  • MDL number MFCD02379956
  • Cas Number : 13289-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=69167

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=69167
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart