APO443485467 Apollo Scientific Ltd

4-Bromo-2,5-difluorobenzaldehyde

  • MDL number MFCD11847047
  • Cas Number : 357405-75-5
  • Molecular Weight : 221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-25-difluorobenzaldehyde

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-25-difluorobenzaldehyde
pictograms GHS06
hstatements H301;H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart