APO443485695 Apollo Scientific Ltd

Methyl 3-bromo-4-(trifluoromethyl)benzoate

  • MDL number MFCD14697998
  • Cas Number : 455941-82-9
  • Molecular Weight : 283.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromo-4-trifluoromethylbenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromo-4-trifluoromethylbenzoate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart