APO443485709 Apollo Scientific Ltd

Ethyl 6-bromo-1-benzofuran-2-carboxylate

  • MDL number MFCD16038162
  • Cas Number : 907945-62-4
  • Molecular Weight : 269.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 68-70
Iupac Name ethyl 6-bromobenzofuran-2-carboxylate
Inchi Code 1S/C11H9BrO3/c1-2-14-11(13)10-5-7-3-4-8(12)6-9(7)15-10/h3-6H,2H2,1H3
Inchi Key BHQBSMLYZIRFHX-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 68-70
Product Link https://store.apolloscientific.co.uk/product/ethyl-6-bromo-1-benzofuran-2-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-6-bromo-1-benzofuran-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart