APO443486049 Apollo Scientific Ltd

2-Amino-4-bromo-5-(trifluoromethyl)phenylamine

  • MDL number MFCD09263998
  • Cas Number : 157590-60-8
  • Molecular Weight : 255.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-bromo-5-trifluoromethylphenylamine

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-bromo-5-trifluoromethylphenylamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart