APO444780032 Apollo Scientific Ltd

2,6-Diazaspiro[3.5]nonane-6-carboxylic acid, 1,1-dimethylethyl ester, hydrochloride

  • MDL number MFCD22380770
  • Cas Number : 1279844-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=71058

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=71058
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart