APO444780199 Apollo Scientific Ltd

2,6-Dichloro-4-methoxypyridine

  • MDL number MFCD00955755
  • Cas Number : 17228-75-0
  • Molecular Weight : 178.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-111
Purity 99
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-4-methoxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-4-methoxypyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart