APO444780225 Apollo Scientific Ltd

8-Acetyl-7-hydroxy-4-methylcoumarin

  • MDL number MFCD00100602
  • Cas Number : 2555-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/8-acetyl-7-hydroxy-4-methylcoumarin

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8-acetyl-7-hydroxy-4-methylcoumarin
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart