APO444780378 Apollo Scientific Ltd

2,6-Dibromo-4-methoxypyridine

  • MDL number MFCD11036227
  • Cas Number : 117873-72-0
  • Molecular Weight : 266.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-139
Purity 99
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dibromo-4-methoxypyridine

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/26-dibromo-4-methoxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart