APO444780389 Apollo Scientific Ltd

5-(Aminomethyl)-2-bromopyridine

  • MDL number MFCD06738744
  • Cas Number : 120740-10-5
  • Molecular Weight : 187.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-aminomethyl-2-bromopyridine

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/5-aminomethyl-2-bromopyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart