APO444780445 Apollo Scientific Ltd

2-Amino-4-methyl-5-pyridinecarboxylic acid

  • MDL number MFCD09835242
  • Cas Number : 179555-11-4
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 270-272
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-methyl-5-pyridinecarboxylic-acid

Safety Information

signalword Warning
pcodes P280;P302+P352
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-methyl-5-pyridinecarboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart