APO444780508 Apollo Scientific Ltd

3-Amino-5-bromo-2-nitropyridine

  • MDL number MFCD11040269
  • Cas Number : 433226-05-2
  • Molecular Weight : 218.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-nitro-3-pyridinamine
Inchi Code 1S/C5H4BrN3O2/c6-3-1-4(7)5(8-2-3)9(10)11/h1-2H,7H2
Inchi Key GOXZJSIRAHNQQG-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-nitropyridin-3-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-5-bromo-2-nitropyridine

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-5-bromo-2-nitropyridine
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart