APO444780514 Apollo Scientific Ltd

Imidazo[1,2-a]pyridine-5-carboxylic acid

  • MDL number MFCD09834780
  • Cas Number : 479028-72-3
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name imidazo[1,2-a]pyridine-5-carboxylic acid
Inchi Code 1S/C8H6N2O2/c11-8(12)6-2-1-3-7-9-4-5-10(6)7/h1-5H,(H,11,12)
Inchi Key IAXRYEXXAPLEGT-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/imidazo12-apyridine-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/imidazo12-apyridine-5-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart