APO444780763 Apollo Scientific Ltd

6-Bromo-3,4-methylenedioxybenzoic acid

  • MDL number MFCD00156994
  • Cas Number : 60546-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=70052

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=70052
Availability details for
Quantity
  • Availability
  • Add to Cart