APO444781023 Apollo Scientific Ltd

2-Chloro-4-methyl-pyridine-3-sulphonyl chloride

  • Cas Number : 1208081-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=70638

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=70638
Availability details for
Quantity
  • Availability
  • Add to Cart