APO444781231 Apollo Scientific Ltd

6-Bromo-2H-isoquinolin-1-one

  • MDL number MFCD07374394
  • Cas Number : 82827-09-6
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-2h-isoquinolin-1-one

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/6-bromo-2h-isoquinolin-1-one
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart