APO444781501 Apollo Scientific Ltd

Pyridinium p-toluenesulfonate

  • MDL number MFCD00013108
  • Cas Number : 24057-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=71617

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=71617
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart