APO444781575 Apollo Scientific Ltd

(1-(tert-Butoxycarbonyl)-1H-indol-3-yl)boronic acid

  • MDL number MFCD06656257
  • Cas Number : 181365-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=71692

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=71692
Availability details for
Quantity
  • Availability
  • Add to Cart