APO444781744 Apollo Scientific Ltd

2-Bromo-4-methoxy-6-nitroaniline

  • MDL number MFCD18088521
  • Cas Number : 10172-35-7
  • Molecular Weight : 247.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-methoxy-6-nitroaniline

Safety Information

signalword Warning
pcodes P280;P271;P261
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-methoxy-6-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart