APO444781810 Apollo Scientific Ltd

3-(Benzyloxy)benzyl bromide

  • MDL number MFCD01318307
  • Cas Number : 1700-31-8
  • Molecular Weight : 277.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 54-56
Iupac Name benzyl 3-(bromomethyl)phenyl ether
Inchi Code 1S/C14H13BrO/c15-10-13-7-4-8-14(9-13)16-11-12-5-2-1-3-6-12/h1-9H,10-11H2
Inchi Key ITJWNXBZSFIJTP-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 54-56
Product Link https://store.apolloscientific.co.uk/product/3-benzyloxybenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-benzyloxybenzyl-bromide
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart