APO444781858 Apollo Scientific Ltd

2-Bromo-3-iodobenzyl alcohol

  • MDL number MFCD11520410
  • Cas Number : 1261644-21-6
  • Molecular Weight : 312.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromo-3-iodophenyl)methanol
Inchi Code 1S/C7H6BrIO/c8-7-5(4-10)2-1-3-6(7)9/h1-3,10H,4H2
Inchi Key PRQUJNCOMDYCMC-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-iodobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-iodobenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart