APO444781859 Apollo Scientific Ltd

2-Bromo-3-iodobenzyl bromide

  • MDL number MFCD12025461
  • Cas Number : 1261649-03-9
  • Molecular Weight : 375.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-iodobenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-iodobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart