APO444782092 Apollo Scientific Ltd

2-Bromo-5-iodo-4-nitroanisole

  • MDL number MFCD28348435
  • Cas Number : 1361021-39-7
  • Molecular Weight : 357.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-4-iodo-2-methoxy-5-nitrobenzene
Inchi Code 1S/C7H5BrINO3/c1-13-7-3-5(9)6(10(11)12)2-4(7)8/h2-3H,1H3
Inchi Key LIQQFDPRYUYNMP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-iodo-4-nitroanisole

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-iodo-4-nitroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart