APO444782093 Apollo Scientific Ltd

5-Bromo-2-iodo-4-methoxyaniline

  • MDL number MFCD28348436
  • Cas Number : 1361021-26-2
  • Molecular Weight : 327.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-iodo-4-methoxyaniline
Inchi Code 1S/C7H7BrINO/c1-11-7-3-5(9)6(10)2-4(7)8/h2-3H,10H2,1H3
Inchi Key FUDQBDFQAXUJLS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-iodo-4-methoxyaniline

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-iodo-4-methoxyaniline
pictograms GHS06
hstatements H301+H311;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart