APO444782335 Apollo Scientific Ltd

tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate

  • MDL number MFCD16250751
  • Cas Number : 1209780-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/tert-butyl-33-difluoro-4-hydroxypiperidine-1-carboxylate

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/tert-butyl-33-difluoro-4-hydroxypiperidine-1-carboxylate
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart