APO444782567 Apollo Scientific Ltd

1,1,1-Trifluoro-N-phenylmethanesulphonamide

  • MDL number MFCD00521820
  • Cas Number : 456-64-4
  • Molecular Weight : 225.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/111-trifluoro-n-phenylmethanesulphonamide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/111-trifluoro-n-phenylmethanesulphonamide
pictograms GHS06
hstatements H301+H311;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart