APO444782659 Apollo Scientific Ltd

2-Fluoro-3-nitrobenzyl bromide

  • MDL number MFCD11110170
  • Cas Number : 946125-65-1
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(bromomethyl)-2-fluoro-3-nitrobenzene
Inchi Code 1S/C7H5BrFNO2/c8-4-5-2-1-3-6(7(5)9)10(11)12/h1-3H,4H2
Inchi Key MIESICVJNXXYKC-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-nitrobenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-nitrobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart