APO444782691 Apollo Scientific Ltd

5-Bromo-3-fluoro-2-nitroanisole

  • MDL number MFCD20527806
  • Cas Number : 1137869-91-0
  • Molecular Weight : 250.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-fluoro-2-nitroanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-fluoro-2-nitroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart