APO444782749 Apollo Scientific Ltd

5-Bromo-3-fluoro-2-iodophenol

  • MDL number MFCD27756663
  • Cas Number : 1805500-75-7
  • Molecular Weight : 316.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-fluoro-2-iodophenol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-fluoro-2-iodophenol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart