APO444782797 Apollo Scientific Ltd

1-Bromo-5-fluoro-2-iodo-3-(trifluoromethyl)benzene

  • MDL number MFCD09800680
  • Cas Number : 1027511-93-8
  • Molecular Weight : 368.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-5-fluoro-2-iodo-3-(trifluoromethyl)benzene
Inchi Code 1S/C7H2BrF4I/c8-5-2-3(9)1-4(6(5)13)7(10,11)12/h1-2H
Inchi Key AXNJIWJQGUCDQF-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-bromo-5-fluoro-2-iodo-3-trifluoromethylbenzene

Safety Information

signalword Warning
pcodes P280;P302+P352
MSDS https://store.apolloscientific.co.uk/product/1-bromo-5-fluoro-2-iodo-3-trifluoromethylbenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart