APO444782806 Apollo Scientific Ltd

4-Bromo-3-fluorophenacyl bromide 250mg pack

  • MDL number MFCD11847180
  • Cas Number : 1003879-02-4
  • Molecular Weight : 295.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-fluorophenacyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart