APO444782807 Apollo Scientific Ltd

4-Bromo-3-fluorophenacylamine hydrochloride

  • MDL number MFCD11847050
  • Cas Number : 1260679-52-4
  • Molecular Weight : 268.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-1-(4-bromo-3-fluorophenyl)ethanone hydrochloride
Inchi Code 1S/C8H7BrFNO.ClH/c9-6-2-1-5(3-7(6)10)8(12)4-11;/h1-3H,4,11H2;1H
Inchi Key SDDJUSCOXLXSAB-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-fluorophenacylamine-hydrochloride

Safety Information

signalword Warning
pcodes P302+P352;P305+P351+P338;P362+P364
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-fluorophenacylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart