APO444782858 Apollo Scientific Ltd

2-Fluoro-4-(methylsulphonyl)benzaldehyde

  • MDL number MFCD13191594
  • Cas Number : 1197193-11-5
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-methylsulphonylbenzaldehyde

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-methylsulphonylbenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart