APO444782893 Apollo Scientific Ltd

4-Bromo-5-methoxy-2-nitroaniline

  • MDL number MFCD13659425
  • Cas Number : 173312-36-2
  • Molecular Weight : 247.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-methoxy-2-nitrophenylamine
Inchi Code 1S/C7H7BrN2O3/c1-13-7-3-5(9)6(10(11)12)2-4(7)8/h2-3H,9H2,1H3
Inchi Key JYXJPWNTXBYKMD-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-5-methoxy-2-nitroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-5-methoxy-2-nitroaniline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart