APO447397539 Apollo Scientific Ltd

(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine

  • MDL number MFCD03411208
  • Cas Number : 53152-69-5
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1r2r-1n1n2n2n-tetramethylcyclohexane-12-diamine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1r2r-1n1n2n2n-tetramethylcyclohexane-12-diamine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart