APO447397542 Apollo Scientific Ltd

(2S)-2-(4-Butylphenyl)propanoic acid

  • MDL number MFCD11053546
  • Cas Number : 404354-76-3
  • Molecular Weight : 206.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2s-2-4-butylphenylpropanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2s-2-4-butylphenylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart