APO447397545 Apollo Scientific Ltd

Methyl (2S)-piperidinecarboxylate hydrochloride

  • MDL number MFCD00672366
  • Cas Number : 18650-39-0
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-177
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2s-piperidinecarboxylate-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2s-piperidinecarboxylate-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart