APO447397550 Apollo Scientific Ltd

1-(Pyridin-3-yl)piperazine dihydrobromide

  • MDL number MFCD11841224
  • Cas Number : 1185369-72-5
  • Molecular Weight : 325.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-pyridin-3-ylpiperazine-dihydrobromide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-pyridin-3-ylpiperazine-dihydrobromide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart