APO447397566 Apollo Scientific Ltd

4-Chloro-2-iodotoluene

  • MDL number MFCD00060661
  • Cas Number : 33184-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.82
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 138
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72345

Safety Information

signalword Warning
pcodes P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72345
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart