APO447397581 Apollo Scientific Ltd

1-Bromo-2-iodoethane

  • MDL number MFCD00041542
  • Cas Number : 590-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 28-32
Physical Form Solid-Low Melt
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 163
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72361

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72361
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart