APO447397586 Apollo Scientific Ltd

4-Chloro-3-iodobenzonitrile

  • MDL number MFCD09025674
  • Cas Number : 914106-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-93
Purity 98
Storage temp. TBC
Physical Form Solid
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72366

Safety Information

signalword Warning
pcodes P304;P340;P280;P301;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72366
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart