APO447397726 Apollo Scientific Ltd

(1,1-Difluoroethyl)benzene

  • MDL number MFCD11975315
  • Cas Number : 657-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 67/47mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72477

Safety Information

signalword Warning
pcodes P210;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72477
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart