APO447397776 Apollo Scientific Ltd

1,3,3,4,4,5,5-Heptafluoro-2-methoxycyclopent-1-ene

  • MDL number MFCD01735620
  • Cas Number : 1759-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 104
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72527

Safety Information

signalword Warning
pcodes P210;P241;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72527
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart